Merck
  • Targeting Pancreatic Cancer Metastasis by Inhibition of Vav1, a Driver of Tumor Cell Invasion.
Targeting Pancreatic Cancer Metastasis by Inhibition of Vav1, a Driver of Tumor Cell Invasion.
Cancer research (2015-05-16)
Gina L Razidlo, Christopher Magnine, Arthur C Sletten, Rachel M Hurley, Luciana L Almada, Martin E Fernandez-Zapico, Baoan Ji, Mark A McNiven
ABSTRACT
Pancreatic cancer, one of the most lethal forms of human cancer, is largely resistant to many conventional chemotherapeutic agents. Although many therapeutic approaches focus on tumor growth, metastasis is a primary factor contributing to lethality. Therefore, novel therapies to target metastatic invasion could prevent tumor spread and recurrence resulting from local and distant metastasis. The protein Vav1 is aberrantly expressed in more than half of pancreatic cancers. Its expression promotes activation of Rac and Cdc42 and leads to enhanced invasion and migration, as well as increased tumor cell survival and proliferation, suggesting that Vav1 could be a potent therapeutic target for pancreatic cancer. The purine analogue azathioprine, well known for its function as an anti-inflammatory compound, was recently shown to function by inhibiting Vav1 signaling in immune cells. We therefore hypothesized that azathioprine could also inhibit Vav1 in pancreatic tumor cells to reduce its proinvasive functions. Indeed, we have found that treatment of cultured pancreatic tumor cells with azathioprine inhibited Vav1-dependent invasive cell migration and matrix degradation, through inhibition of Rac and Cdc42 signaling. Furthermore, azathioprine treatment decreased metastasis in both xenograft and genetic mouse models of pancreatic cancer. Strikingly, metastasis was dramatically reduced in Vav1-expressing tumors arising from p48(Cre/+), Kras(G12D/+), p53(F/+) mice. These inhibitory effects were mediated through Vav1, as Vav1-negative cell lines and tumors were largely resistant to azathioprine treatment. These findings demonstrate that azathioprine and related compounds could be potent antimetastatic agents for Vav1-positive pancreatic tumors.
Sigma-Aldrich
Glycerol, SAJ first grade, ≥98.0%
Sigma-Aldrich
Glycerol, JIS special grade, ≥99.0%
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Glycerol, BioUltra, for molecular biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Sodium fluoride, BioXtra, ≥99%
Sigma-Aldrich
Sodium fluoride, BioReagent, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Anti-Actin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Glycerol, for molecular biology, ≥99.0%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, SAJ special grade, ≥99.0%
Sigma-Aldrich
Sodium fluoride, JIS special grade, ≥99.0%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Sodium fluoride, 99.99% trace metals basis
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)