Merck
  • Photochemical properties and phototoxicity of Pazufloxacin: a stable and transient study.
Photochemical properties and phototoxicity of Pazufloxacin: a stable and transient study.
Journal of photochemistry and photobiology. B, Biology (2012-12-12)
Yancheng Liu, Peng Zhang, Haixia Li, Ruizhi Tang, Rong rong Cui, Wenfeng Wang
ABSTRACT
Photochemical properties and phototoxicity of Pazufloxacin (PAX) were systematically investigated in aqueous solutions using UV-Vis, fluorescence, laser flash photolysis, pulse radiolysis and SDS-PAGE gel electrophoresis techniques. PAX triplet-state ((3)PAX(*)) absorption spectra (λ(max)=570 nm) was determined. (3)PAX(*) was quenched by PAX and O(2), with rate constants of 6.9×10(8) and 3.2×10(8) dm(3) mol(-1) s(-1), respectively. The pK(a) values (5.7 and 8.6) for the protonation equilibrium were determined by UV-Vis and fluorescence techniques. The PAX triplet energy (E(T)=260.3 kJ/mol) was obtained using energy transfer method. The reaction of electron transfer from tryptophan (TrpH) and dGMP to (3)PAX(*) was found with rate constants of 8.8×10(7) and 8.7×10(6) dm(3) mol(-1) s(-1), respectively. The rate constants for reactions of ()OH, SO(4)(-) and hydrated electron with PAX were found to be 5.8×10(8), 2.1×10(9) and 9×10(9)d m(3) mol(-1) s(-1), respectively. Based on the results obtained, a rational scheme for dGMP, TrpH and lysozyme photodamage induced by PAX was proposed.
Sigma-Aldrich
2′-Deoxyguanosine 5′-monophosphate disodium salt hydrate, ≥98%