Merck
  • A hybrid neutron diffraction and computer simulation study on the solvation of N-methylformamide in dimethylsulfoxide.
A hybrid neutron diffraction and computer simulation study on the solvation of N-methylformamide in dimethylsulfoxide.
The Journal of chemical physics (2013-02-08)
João M M Cordeiro, Alan K Soper
ABSTRACT
The solvation of N-methylformamide (NMF) by dimethylsulfoxide (DMSO) in a 20% NMF/DMSO liquid mixture is investigated using a combination of neutron diffraction augmented with isotopic substitution and Monte Carlo simulations. The aim is to investigate the solute-solvent interactions and the structure of the solution. The results point to the formation of a hydrogen bond (H-bond) between the H bonded to the N of the amine group of NMF and the O of DMSO particularly strong when compared with other H-bonded liquids. Moreover, a second cooperative H-bond is identified with the S atom of DMSO. As a consequence of these H-bonds, molecules of NMF and DMSO are rather rigidly connected, establishing very stable dimmers in the mixture and very well organized first and second solvation shells.
Sigma-Aldrich
N-Methylformamide, 99%