- The selective treatment of clinical mastitis based on on-farm culture results: II. Effects on lactation performance, including clinical mastitis recurrence, somatic cell count, milk production, and cow survival.
The selective treatment of clinical mastitis based on on-farm culture results: II. Effects on lactation performance, including clinical mastitis recurrence, somatic cell count, milk production, and cow survival.
The objective of this multi-state, multi-herd clinical trial was to report on the efficacy of using an on-farm culture system to guide strategic treatment decisions in cows with clinical mastitis. The study was conducted in 8 commercial dairy farms ranging in size from 144 to 1,795 cows from Minnesota, Wisconsin, and Ontario, Canada. A total of 422 cows affected with mild or moderate clinical mastitis in 449 quarters were randomly assigned to either (1) a positive-control treatment program or (2) an on-farm culture-based treatment program. Quarter cases assigned to the positive-control group received immediate on-label intramammary treatment with cephapirin sodium. Quarters assigned to the culture-based treatment program were not treated until the results of on-farm culture were determined after 18 to 24h of incubation. Quarters in the culture-based treatment program that had gram-positive growth or a mixed infection were treated according to label instruction using intramammary cephapirin sodium. Quarters assigned to the culture-based treatment program that had gram-negative or no-growth did not receive intramammary therapy. It was already reported in a companion paper that the selective treatment of clinical mastitis based on on-farm culture results decreases antibiotic use by half and tends to decrease milk withholding time without affecting short-term clinical and bacteriological outcomes. The present article reports on long-term outcomes of the aforementioned study. No statistically significant differences existed between cases assigned to the positive-control program and cases assigned to the culture-based treatment program in risk and days for recurrence of clinical mastitis in the same quarter (35% and 78 d vs. 43% and 82 d), linear somatic cell count (4.2 vs. 4.4), daily milk production (30.0 vs. 30.7 kg), and risk and days for culling or death events (28% and 160 d vs. 32% and 137 d) for the rest of the lactation after enrollment of the clinical mastitis case. In summary, the selective treatment of clinical mastitis based on on-farm culture resulted in no differences in long-term outcomes, such as recurrence of clinical mastitis in the same quarter, somatic cell count, milk production, and cow survival for the rest of the lactation after clinical mastitis.