Merck
  • Supramolecular construction of vesicles based on core-substituted naphthalene diimide appended with triethyleneglycol motifs.
Supramolecular construction of vesicles based on core-substituted naphthalene diimide appended with triethyleneglycol motifs.
Chemical communications (Cambridge, England) (2011-05-10)
Sheshanath V Bhosale, Chintan H Jani, Cecilia H Lalander, Steven J Langford, Igor Nerush, Joseph G Shapter, Diego Villamaina, Eric Vauthey
ABSTRACT
The self-assembly of core-substituted naphthalene diimides bearing triethylene glycol motifs leads to the formation of stable vesicles in DMSO and CHCl(3)/MeOH (6 : 4, v/v) solvents. The vesicles were evaluated by means of UV/vis and fluorescence spectroscopy, transmission electron microscopy, atomic force microscopy and dynamic light scattering.
Sigma-Aldrich
Triethylene glycol, ReagentPlus®, 99%
Sigma-Aldrich
Triethylene glycol, BioUltra, anhydrous, ≥99.0% (GC)