Merck
  • Chromium improves glucose uptake and metabolism through upregulating the mRNA levels of IR, GLUT4, GS, and UCP3 in skeletal muscle cells.
Chromium improves glucose uptake and metabolism through upregulating the mRNA levels of IR, GLUT4, GS, and UCP3 in skeletal muscle cells.
Biological trace element research (2009-03-14)
Wei Qiao, Zhongli Peng, Zhisheng Wang, Jing Wei, Anguo Zhou
ABSTRACT
The aim of this study was to evaluate the impact of three different chromium forms as chromic chloride (CrCl), chromium picolinate (CrPic), and a newly synthesized complex of chromium chelated with small peptides (CrSP) on glucose uptake and metabolism in vitro. In cultured skeletal muscle cells, chromium augmented insulin-stimulated glucose uptake and metabolism as assessed by a reduced glucose concentration of culture medium. At the molecular level, insulin significantly increased the mRNA levels of insulin receptor (IR), glucose transporter 4 (GLUT4), glycogen synthase (GS), and uncoupling protein-3 (UCP3), and these impacts can be enhanced by the addition of chromium, especially in the form of CrSP. Collectively, results of this study demonstrate that chromium improves glucose uptake and metabolism through upregulating the mRNA levels of IR, GLUT4, GS, and UCP3 in skeletal muscle cells, and CrSP has higher efficacy on glucose uptake and metabolism compared to the forms of CrCl and CrPic.
Sigma-Aldrich
Chromium(III) chloride, anhydrous, 99.99% trace metals basis
Sigma-Aldrich
Chromium(III) chloride, purified by sublimation, 99%
Sigma-Aldrich
Chromium picolinate, ≥98% (HPLC)