Merck
  • The PSI-E subunit of photosystem I binds ferredoxin:NADP+ oxidoreductase.
The PSI-E subunit of photosystem I binds ferredoxin:NADP+ oxidoreductase.
FEBS letters (1992-10-19)
B Andersen, H V Scheller, B L Møller
ABSTRACT
A photosystem I complex containing the polypeptides PSI-A to PSI-L, light-harvesting complex I and ferredoxin:NADP+ oxidoreductase has been isolated from barley using the non-ionic detergent n-decyl-beta-D-maltopyranoside. The ratio between bound ferredoxin:NADP+ oxidoreductase and P700 is 0.4 +/- 0.2. The complex is highly active in catalyzing light-induced transfer of electrons from plastocyanin to NADP+ at rates of 280 +/- 150 and 1800 +/- 800 mumol NADPH/(mg chl.h), without and in the presence of saturating amounts of exogenously added ferredoxin:NADP+ oxidoreductase, respectively. Endogenously bound ferredoxin:NADP+ oxidoreductase interacts with the PSI-E subunit as demonstrated by cross-linking experiments using two different types of cross-linkers and identification of the products by Western blotting and the use of monospecific antibodies.
Sigma-Aldrich
Decyl β-D-maltopyranoside, ≥98% (GC)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPrice
100 mg
1 Estimated to ship on 09 March 2025
MYR 117.00
1 g
1 Estimated to ship on 09 March 2025
MYR 466.00
5 x 1 g
1 Estimated to ship on 09 March 2025
MYR 2,311.00
To order products, please contact your local dealer.