
Triolein

Synonym(s):
Glyceryl trioleate, (9Z)9-Octadecenoic acid 1,2,3-propanetriyl ester, 1,2,3-Tri(cis-9-octadecenoyl)glycerol, Glycerol trioleate, Glycerol triolein, Oleic acid triglyceride, Oleic triglyceride, TG(18:1(9Z)/18:1(9Z)/18:1(9Z)), Triolein
Linear Formula:
(C17H33COOCH2)2CHOCOC17H33
CAS Number:
Molecular Weight:
885.43
Products
Boiling Point (°C)
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing