
Quisqualic acid

Synonym(s):
β-(3,5-Dioxo-1,2,4-oxadiazolidin-2-yl)-L-alanine, 3-(3,5-Dioxo-1,2,4-oxadiazolidin-2-yl)-L-alanine
Empirical Formula (Hill Notation):
C5H7N3O5
CAS Number:
Molecular Weight:
189.13
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Physical Form
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing