
ORBO™ 34 Activated Coconut Charcoal specially treated (20/40), 400/200 mg

Synonym(s):
Activated Coconut Charcoal Tube
Products
Brand
Product Line
Agency
Chemical Composition
OD (mm)
Quality Segment
Application
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing