
L-Fmoc-4-fluorophenylalanine

Synonym(s):
Fmoc-Phe(4-F)-OH, Fmoc-4-fluoro-L-phenylalanine
Empirical Formula (Hill Notation):
C24H20FNO4
CAS Number:
Molecular Weight:
405.42
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Amino Acid Type
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing