
Ketamine impurity A

Synonym(s):
1-[(2-Chlorophenyl)(methylimino)methyl]cyclopentanol
Empirical Formula (Hill Notation):
C13H16ClNO
CAS Number:
Molecular Weight:
237.73
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing