
Ephedrine hydrochloride (racemic)

Synonym(s):
DL-Ephedrine hydrochloride
Linear Formula:
C6H5CH(OH)CH(CH3)NHCH3 · HCl
CAS Number:
Molecular Weight:
201.69
Products
Formula Weight
Manufacturer
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing