![Amyloid β Protein Fragment 1-40 ≥90% (HPLC), powder](/deepweb/assets/sigmaaldrich/product/images/302/805/6d5ade41-da2c-4381-bcf3-476e964c4c30/640/6d5ade41-da2c-4381-bcf3-476e964c4c30.jpg)
Amyloid β Protein Fragment 1-40
![Amyloid β Protein Fragment 1-40 ≥90% (HPLC), powder](/deepweb/assets/sigmaaldrich/product/images/302/805/6d5ade41-da2c-4381-bcf3-476e964c4c30/640/6d5ade41-da2c-4381-bcf3-476e964c4c30.jpg)
Synonym(s):
Aβ40
Empirical Formula (Hill Notation):
C194H295N53O58S
CAS Number:
Molecular Weight:
4329.80
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing