Skip to Content
Merck
All Photos(1)

Key Documents

SRP6084

Sigma-Aldrich

ALDH3A1 human

recombinant, expressed in E. coli, ≥95% (SDS-PAGE)

Synonym(s):

ALDH3, ALDHIII, Aldehyde dehydrogenase 3, Aldehyde dehydrogenase family 3 member A1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352204
NACRES:
NA.77

biological source

human

recombinant

expressed in E. coli

Assay

≥95% (SDS-PAGE)

form

liquid

mol wt

52.5 kDa

packaging

pkg of 50 μg

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... ALDH3A1(218)

General description

ALDH3A1, also known as Aldehyde dehydrogenase 3 family memberA1, is involved in the detoxification of alcohol-derived acetaldehyde and in the metabolism of corticosteroids, biogenic amines, neurotransmitters, and lipid peroxidation. This protein forms a cytoplasmic homodimer that preferentially oxidizes aromatic and medium-chain (6 carbons or more) saturated and unsaturated aldehyde substrates. It is thought to promote resistance to UV and 4-hydroxy-2-nonenal-induced oxidative damage in the cornea.

Physical form

0.5 mg/mL solution in 20 mM Tris-HCl buffer (pH 8.0) containing 0.1 M NaCl and 10% glycerol.

Preparation Note

Centrifuge the vial prior to opening.

Other Notes

MGSSHHHHHH SSGLVPRGSH MSKISEAVKR ARAAFSSGRT RPLQFRIQQL EALQRLIQEQ EQELVGALAA DLHKNEWNAY YEEVVYVLEE IEYMIQKLPE WAADEPVEKT PQTQQDELYI HSEPLGVVLV IGTWNYPFNL TIQPMVGAIA AGNAVVLKPS ELSENMASLL ATIIPQYLDK DLYPVINGGV PETTELLKER FDHILYTGST GVGKIIMTAA AKHLTPVTLE LGGKSPCYVD KNCDLDVACR RIAWGKFMNS GQTCVAPDYI LCDPSIQNQI VEKLKKSLKE FYGEDAKKSR DYGRIISARH FQRVMGLIEG QKVAYGGTGD AATRYIAPTI LTDVDPQSPV MQEEIFGPVL PIVCVRSLEE AIQFINQREK PLALYMFSSN DKVIKKMIAE TSSGGVAAND VIVHITLHSL PFGGVGNSGM GSYHGKKSFE TFSHRRSCLV RPLMNDEGLK VRYPPSPAKM TQH

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service