HPLC Analysis of α-Bromopalmitic Acid Enantiomers on Astec® CHIROBIOTIC® T2
CONDITIONS
column
CHIROBIOTIC T2, 25 cm x 4.6 mm I.D., 5 μm particles (16024AST)
mobile phase
[A] 20 mM ammonium acetate, pH 5.5; [B] methanol; (40:60, A:B)
flow rate
0.9 mL/min
column temp.
25 °C
detector
UV, 230 nm
injection
5 μL
sample
2 mg/mL in methanol
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC