Merck
  • LC/ESI-MS/MS method for determination of salivary eicosapentaenoic acid concentration to arachidonic acid concentration ratio.
LC/ESI-MS/MS method for determination of salivary eicosapentaenoic acid concentration to arachidonic acid concentration ratio.
Biomedical chromatography : BMC (2015-01-27)
Shoujiro Ogawa, Koki Tomaru, Nagisa Matsumoto, Shui Watanabe, Tatsuya Higashi
ABSTRACT
A simple liquid chromatography/electrospray ionization-tandem mass spectrometry (LC/ESI-MS/MS) method for determination of the eicosapentaenoic acid (EPA) concentration to arachidonic acid (AA) concentration ratio in human saliva has been developed. The EPA/AA ratio in serum or plasma is widely recognized as a useful indicator in identifying the risk of cardiovascular disease, especially atherosclerosis. The salivary EPA/AA ratio is expected to be a convenient alternative to the serum or plasma EPA/AA ratio, because saliva offers the advantages of easy and noninvasive sampling. The saliva was deproteinized with acetonitrile, purified using an Oasis HLB cartridge, and derivatized with 1-[(4-dimethylaminophenyl)carbonyl]piperazine (DAPPZ). The derivatized EPA and AA were subjected to LC/ESI-MS/MS, and the EPA/AA ratio was determined using the selected reaction monitoring mode. The DAPPZ-derivatization increased the ESI sensitivity by 100- and 300-fold for EPA and AA, respectively, and enabled the detection of trace fatty acids in saliva using a 200 μL sample. The assay reproducibility was satisfactory (relative standard deviation, <5.0%). The method was successfully applied to the measurement of the salivary EPA/AA ratios of healthy Japanese subjects and their changes owing to the supplementation of EPA.
Sigma-Aldrich
(±)-2-(p-Methoxyphenoxy)propionic acid, ≥98%
Sigma-Aldrich
(S)-(+)-1-(2-Pyrrolidinylmethyl)pyrrolidine, 96%