Merck
  • Sequential and one-pot reactions of phenols with bromoalkynes for the synthesis of (Z)-2-bromovinyl phenyl ethers and benzo[b]furans.
Sequential and one-pot reactions of phenols with bromoalkynes for the synthesis of (Z)-2-bromovinyl phenyl ethers and benzo[b]furans.
Organic letters (2011-10-20)
Shihua Wang, Pinhua Li, Lin Yu, Lei Wang
ABSTRACT
Benzo[b]furans were prepared in one pot based on the addition/palladium-catalyzed C-H bond functionalization of phenols with bromoalkynes. The addition reactions of phenols to bromoalkynes generated (Z)-2-bromovinyl phenyl ethers in high yields with excellent regio- and stereoselectivity. The obtained (Z)-2-bromovinyl phenyl ethers subsequently proceeded by intramolecular cyclization to afford 2-substituted benzo[b]furans in good yields through palladium-catalyzed direct C-H bond functionalizations. It is important to note that the transformation of phenols with bromoalkynes into benzo[b]furans could be carried out in one pot with a simple and efficient tandem procedure.
Sigma-Aldrich
Phenyl vinyl ether, 97%