Merck
  • The molecular basis of host plant selection in Melaleuca quinquenervia by a successful biological control agent.
The molecular basis of host plant selection in Melaleuca quinquenervia by a successful biological control agent.
Phytochemistry (2010-06-18)
Amanda Padovan, Andras Keszei, Tobias G Köllner, Jörg Degenhardt, William J Foley
ABSTRACT
Melaleuca quinquenervia (Cav) S.T. Blake (broadleaf paperbark) is an Australian tree that has become a serious weed in many places around the world. Two insects Oxyops vitiosa (the melaleuca weevil), and Boreioglycaspis melaleucae (the melaleuca psyllid), which were introduced to Florida as part of a biological control programme, have been very effective in reducing survival and reproduction of this weed. There are two terpene chemotypes of M. quinquenervia; one rich in the sesquiterpene E-nerolidol whereas the other is rich in viridiflorol. Viridiflorol is a strong feeding deterrent for the melaleuca weevil and retards larval development. The larvae therefore avoid the viridiflorol-rich chemotype, in contrast, female melaleuca psyllids prefer to oviposit on these leaves. To identify the molecular basis of these preferences, we isolated and characterised two terpene synthases from the viridiflorol-rich chemotype, both of which utilise farnesyl pyrophosphate and have the same product profile. Chemotypic variation in terpenes in M. quinquenervia is under strong genetic control and the reproductive potential of each chemotype is limited by a different insect. These insects could, therefore, be selective agents for the maintenance of chemotypic variation in M. quinquenervia.
Sigma-Aldrich
Nerolidol, 98%, Mixture of cis and trans
Sigma-Aldrich
Nerolidol, mixture of cis and trans, ≥97%, stabilized, FG
Supelco
trans-Nerolidol, analytical standard