Merck
  • Relationship between high sensitivity of suckling mice to hepatocarcinogenic and antiglucocorticoid effects of o-aminoazotoluene and diethylnitrosamine.
Relationship between high sensitivity of suckling mice to hepatocarcinogenic and antiglucocorticoid effects of o-aminoazotoluene and diethylnitrosamine.
Bulletin of experimental biology and medicine (2004-09-30)
S I Il'nitskaya, E D Vasil'eva, K Yu Kropachev, V I Kaledin
ABSTRACT
Suckling mice were more sensitive to the hepatocarcinogenic effect of various carcinogens compared to adult animals. After treatment with o-aminoazotoluene and diethylnitrosamine HNF3-DNA-binding capacity and glucocorticoid-induced liver tyrosine aminotransferase activity in suckling mice decreased more significantly than in adult animals.
Product Description
Supelco
o-Aminoazotoluene, analytical standard
Sigma-Aldrich
Fast Garnet GBC base, 97%