Merck
  • Synthesis, characterization, and alkoxide transfer reactivity of dimeric Tl2(OR)2 complexes.
Synthesis, characterization, and alkoxide transfer reactivity of dimeric Tl2(OR)2 complexes.
Dalton transactions (Cambridge, England : 2003) (2021-01-31)
Amanda Grass, Lakshani Wathsala Kulathungage, Duleeka Wannipurage, Maryam Yousif, Cassandra L Ward, Stanislav Groysman
ABSTRACT
Reaction of LiOCtBu2Ph with TlPF6 forms the dimeric Tl2(OCtBu2Ph)2 complex, a rare example of a homoleptic thallium alkoxide complex demonstrating formally two-coordinate metal centers. Characterization of Tl2(OCtBu2Ph)2 by 1H and 13C NMR spectroscopy and X-ray crystallography reveals the presence of two isomers differing by the mutual conformation of the alkoxide ligands, and by the planarity of the central Tl-O-Tl-O plane. Tl2(OCtBu2Ph)2 serves as a convenient precursor to the formation of old and new [M(OCtBu2Ph)n] complexes (M = Cr, Fe, Cu, Zn), including a rare example of T-shaped Zn(OCtBu2Ph)2(THF) complex, which could not be previously synthesized using more conventional LiOR/HOR precursors. The reaction of [Ru(cymene)Cl2]2 with Tl2(OCtBu2Ph)2 results in the formation of a ruthenium(ii) alkoxide complex. For ruthenium, the initial coordination of the alkoxide triggers C-H activation at the ortho-H of [OCtBu2Ph] which results in its bidentate coordination. In addition to Tl2(OCtBu2Ph)2, related Tl2(OCtBu2(3,5-Me2C6H3))2 was also synthesized, characterized, and shown to exhibit similar reactivity with iron and ruthenium precursors. Synthetic, structural, and spectroscopic characterizations are presented.
Sigma-Aldrich
Nickel(II) chloride ethylene glycol dimethyl ether complex, 98%
Pricing and availability is not currently available.