Merck
  • Pan-cancer analysis reveals synergistic effects of CDK4/6i and PARPi combination treatment in RB-proficient and RB-deficient breast cancer cells.
Pan-cancer analysis reveals synergistic effects of CDK4/6i and PARPi combination treatment in RB-proficient and RB-deficient breast cancer cells.
Cell death & disease (2020-04-07)
Songyu Li, Yixiang Zhang, Na Wang, Rong Guo, Qiaoling Liu, Changsheng Lv, Jinguang Wang, Lina Wang, Qing-Kai Yang
ABSTRACT
DNA damage results in mutations and plays critical roles in cancer development, progression, and treatment. Targeting DNA damage response in cancers by inhibiting poly-(ADP-ribose) polymerases (PARPs) offers an important therapeutic strategy. However, the failure of PARP inhibitors to markedly benefit patients suggests the necessity for developing new strategies to improve their efficacy. Here, we show that the expression of cyclin-dependent kinase 4/6 (CDK4/6) complex members significantly correlates with mutations (as proxies of DNA damages), and that the combination of CDK4/6 and PARP inhibitors shows synergy in both RB-proficient and RB-deficient breast cancer cells. As PARPs constitute sensors of DNA damage and are broadly involved in multiple DNA repair pathways, we hypothesized that the combined inhibition of PARPs and DNA repair (or repair-related) pathways critical for cancer (DRPCC) should show synergy. To identify druggable candidate DRPCC(s), we analyzed the correlation between the genome-wide expression of individual genes and the mutations for 27 different cancer types, assessing 7146 exomes and over 1,500,000 somatic mutations. Pathway enrichment analyses of the top-ranked genes correlated with mutations indicated "cell cycle pathway" as the top candidate DRPCC. Additionally, among functional cell-cycle complexes, the CDK4/6 complex showed the most significant negative correlation with mutations, also suggesting that combined CDK4/6 and PARP inhibition might exhibit synergy. Furthermore, combination treatment showed synergy in not only RB-proficient but also RB-deficient breast cancer cells in a reactive oxygen species-dependent manner. These findings suggest a potential therapeutic strategy to improve the efficacy of PARP and CDK4/6 inhibitors in cancer treatment.
Sigma-Aldrich
Anti-phospho-Histone H2A.X (Ser139) Antibody, clone JBW301, clone JBW301, Upstate®, from mouse
Sigma-Aldrich
Anti-BrdU Antibody, clone PRB-1, FITC conjugated, clone PRB-1, Chemicon®, from mouse