Merck
  • Depletion of ID3 enhances mesenchymal stem cells therapy by targeting BMP4 in Sjögren's syndrome.
Depletion of ID3 enhances mesenchymal stem cells therapy by targeting BMP4 in Sjögren's syndrome.
Cell death & disease (2020-03-07)
Lei Hu, Junji Xu, Tingting Wu, Zhipeng Fan, Lingyun Sun, Yi Liu, Yan Li, Chunmei Zhang, Jingsong Wang, Yaozhong Ding, Songlin Wang
ABSTRACT
Mesenchymal stem cell (MSCs) transplantation has been used to treat Sjögren's syndrome (SS) based on the immunoregulatory properties of MSCs. However, the effectiveness need improving and its underlying intrinsic mechanisms remain largely unknown. Here, we show that Id3 is upregulated in bone marrow-derived MSCs (BMMSCs) isolated from NOD/ShiLtJ mice, a widely used SS model, compared with ICR mice as control, suggesting that it functions in SS development and therapy. Transplantation of Id3-deficient BMMSCs rescues salivary gland function more effective than wild-type BMMSCs in NOD/ShiLtJ mice. Mechanistically, we show that ID3 negatively regulated BMP4 expression by preventing binding of basic helix-loop-helix protein E2A to the promoter of the Bmp4 gene. BMP4 in turn promoted PGE2 production in MSCs, and exhibited enhanced suppressive activities of T-cell proliferation and Th1 differentiation. Importantly, BMMSCs from SS patients showed significantly lower BMP4 and PGE2 expression than those from healthy individuals. Taken together, our findings revealed the targeting Id3 may be therapeutically useful for improving MSC immunoregulation and effectiveness of MSCs therapy for SS.
Sigma-Aldrich
MISSION® esiRNA, targeting human SERPINB2, SERPINB10
Sigma-Aldrich
Pomalidomide, ≥98% (HPLC)
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Lce1h