Merck
  • Determination of the cis-trans isomerization barrier of several L-peptidyl-L-proline dipeptides by dynamic capillary electrophoresis and computer simulation.
Determination of the cis-trans isomerization barrier of several L-peptidyl-L-proline dipeptides by dynamic capillary electrophoresis and computer simulation.
Electrophoresis (2001-08-25)
G Schoetz, O Trapp, V Schurig
ABSTRACT
Dynamic capillary electrophoresis (DCE) and computer simulation of the elution profiles with the theoretical plate and the stochastic model has been applied to determine the isomerization barriers of the three dipeptides L-alanyl-L-proline, L-leucyl-L-proline, and L-phenylalanyl-L-proline. The separation of the rotational cis-trans isomers has been performed in an aqueous 70 mM borate buffer at pH 9.5. Interconversion profiles featuring plateau formation and peak broadening were observed. To determine the rate constants k1 and k(-1) of the cis-trans isomerization in dynamic capillary electrophoresis, equations have been derived for the theoretical plate model and stochastic model. The electropherograms were simulated with the ChromWin software which uses the experimental data plateau height h(plateau), peak width at half height Wh, the total migration times of the cis-trans isomers tR and the electroosmotic break-through time t0 as well as the peak ratio [cis]/[trans]. From temperature-dependent measurements, the rate constants k1 and k(-1) and the kinetic activation parameters deltaG#, deltaH# and deltaS# of the cis-trans isomerization of the three dipeptides were obtained.
Sigma-Aldrich
Leu-Pro hydrochloride
Sigma-Aldrich
Ala-Pro hydrate
Sigma-Aldrich
Phe-Pro