
Tigecycline for system suitability

Synonym(s):
Tigecycline, 9-tert-Butylglycylamidominocycline
Empirical Formula (Hill Notation):
C29H39N5O8
CAS Number:
Molecular Weight:
585.65
Products
Formula Weight
Manufacturer
Format
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing