
S-Kinoprene

Synonym(s):
S-Kinoprene, 2-Propyn-1-yl (2E,4E,7S)-3,7,11-trimethyl-2,4-dodecadienoate
Empirical Formula (Hill Notation):
C18H28O2
CAS Number:
Molecular Weight:
276.41
Products
Brand
Formula Weight
Quality Segment
Application
Package Size
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing