
N-(2-Chloro-4-pyridyl)-N′-phenylurea

Synonym(s):
4-CPPU, CPPU
Empirical Formula (Hill Notation):
C12H10ClN3O
CAS Number:
Molecular Weight:
247.68
Products
Brand
Formula Weight
Physical Form
Product Line
Quality Segment
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing