
Linolenyl alcohol

Synonym(s):
9,12,15-Octadecatrienol
Empirical Formula (Hill Notation):
C18H32O
CAS Number:
Molecular Weight:
264.45
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2