
DMT-2′O-MOE-rMeU Phosphoramidite

Synonym(s):
5′-Dimethoxytrityl-3′-[(2-cyanoethyl)- (N,N-diisopropyl)]-phosphoramidite- 2′-O-(2-methoxyethyl)-5-methyluridine
Empirical Formula (Hill Notation):
C43H55N4O10P
CAS Number:
Molecular Weight:
818.89
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Physical Form
Product Line
Quality Segment
Showing 1-2 of 2
Compare
Product Number
Description
Pricing