
Cyclopropylamine

Synonym(s):
Aminocyclopropane, Cyclopropylamine
Linear Formula:
C3H5NH2
CAS Number:
Molecular Weight:
57.09
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing