
Anti-ADAR2 antibody, Mouse monoclonal

Synonym(s):
Anti-ADARB1, Anti-Adenosine Deaminase, RNA-specific B1, Anti-DRABA2, Anti-DRADA2, Anti-RED1, Anti-RNA Editase 1, Anti-RNA-editing enzyme 1
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Isotype
Physical Form
Antibody Form
Research Area
Conjugate
Package Size
Post Translational Modification
Species Reactivity
Technique
Showing 1-2 of 2