Key Documents
EP5427710000
Eppendorf® Rotor FA-45-24-11-HS
incl. rotor lid, aerosol-tight, coated
About This Item
UNSPSC Code:
41103909
NACRES:
NB.82
feature
to hold 24 x 1.5/2.0 mL tubes
incl. rotor lid, aerosol-tight, coated
packaging
pack of 1 ea
manufacturer/tradename
Eppendorf® 5427710000
parameter
175,000 max. RCF (30,130 xg)
technique(s)
centrifugation: suitable
compatibility
for use with centrifuge models 5430/5430R
Legal Information
Eppendorf is a registered trademark of Eppendorf AG