Documents
CLS6710
Corning® LSE Multiposition Digital Vortexer
AC/DC input 230 V, AC/DC input 50 Hz, AC/DC input, Schuko plug, AC/DC input, UK plug
About This Item
UNSPSC Code:
41103814
NACRES:
NB.02
manufacturer/tradename
Corning 6710
6710
chamber W × D × H
(10.3 in.) (13.3 in.) (5.1 in.)
General description
The Corning Digital Vortexer is a multipurpose shaker dedicated to a variety of shaking and mixing applications. Three easily removable and interchangeable platforms accommodate many different sample containers such as flasks, bottles, tube racks, plates, boxes, and dishes. The clamp platform has spring loaded retaining bars that adjust to fit vessels of different shapes and sizes. The microplate platform is designed to securely hold up to four microplates. Operation of the vortexer can be timed or continuous, while the speed is variable. The broad speed range allows the vortexer to be used for gentle mixing as well as vigorous agitation. A powerful motor, and a chemically resistant and easy to clean surface makes the equipment a perfect choice for years of trouble-free use.
Features and Benefits
  • Three versatile platform options (available separately)
  • Compact design with easy-access control panel
  • Non-slip rubber mat to keep containers in place
  • Safe for cold room or incubator use
Legal Information
Corning is a registered trademark of Corning, Inc.
LSE is a trademark of Corning, Inc.