Skip to Content
Merck
All Photos(1)

Key Documents

AB3843

Sigma-Aldrich

Anti-Ezrin/Radixin/Moesin Antibody

Chemicon®, from rabbit

Synonym(s):

Anti-EBP50, Anti-NHERF, Anti-NHERF-1, Anti-NHERF1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, monkey, rat, human

manufacturer/tradename

Chemicon®

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable (paraffin)
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... SLC9A3R1(9368)

Specificity

Recognizes Ezrin/Radixin/Moesin. These proteins function as linkers between the plasma membrane and the actin cytoskeleton, and are involved in cell adhesion, membrane ruffling, and microvilli formation. This antibody does not cross-react with other related proteins such as merlin or band 4.1.

Immunogen

KLH-conjugated, synthetic peptide corresponding to residues surrounding Thr567 of human ezrin.

Application

Detect Ezrin/Radixin/Moesin using this Anti-Ezrin/Radixin/Moesin Antibody validated for use in WB, IC, IH(P).
Research Category
Cell Structure
Research Sub Category
Adhesion (CAMs)
Western blotting: 1:1,000. The 75 kDa band is Moesin and the 80kDa band is Ezrin and Radixin. (For best results, incubate membrane with diluted antibody in 5% BSA, 1X TBS, 0.1% Tween-20 at 4°C with gentle shaking, overnight)

Immunohistochemistry (Paraffin): 1:50. Positive control tissue is breast carcinoma. Cellular localization is membrane and cytoplasm

Immunocytochemistry (IF): 1:100

Optimal working dilutions must be determined by the end user.

Physical form

Protein A and peptide affinity purified immunoglobulin. Liquid in 10 mM sodium HEPES, pH 7.5, 150 mM NaCl, 100 μg/mL BSA, with 50% glycerol.

Storage and Stability

Maintain at -20ºC for 12 months, do not aliquot.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service