Microbiology Quality Control in Food and Beverage
Food contaminated by microorganisms (bacteria and yeasts), viruses, and protozoa can cause severe disease in humans. There are two categories of foodborne diseases.
- First, food poisoning is caused by the presence of microbial toxins in food products, e.g., by Staphylococcus aureus, Clostridium perfringens (both produce enterotoxins which elicit enteric disease such as diarrhea), and Clostridium botulinum (botulism is the most severe type of food poisoning).
- Second, the growth of microorganisms in the body after eating contaminated food, e.g., by Salmonella spp. (salmonellosis) and Campylobacter jejuni (high fever, abdominal cramps). Many human pathogens are transmitted by fecal-contaminated water, the most important being Salmonella typhi (typhoid fever) and Vibrio cholerae (cholera).
- Microbial Testing Requirements
- Culture Media for Pathogen Detection
- Test for Classic Biochemical Methods
- Test Kits for Innovative Molecular Biological Methods
- Test Kits for Innovative Immunological Methods
Microbial Testing Requirements
Microbiological test procedures for the examination of foods and beverages have been standardized and regulated, but nearly every country has its own regulations. Most of the culture media, additives, dyes, and indicators conform to standard norms like ISO, these products include:
- Culture media and base ingredients
- Chromogenic media for differentiation of microbes based on colony color
- Test kits (biochemical, immunological, and molecular biological methods) for pathogen detection, identification, and confirmation
- Vitroid certified reference microorganisms to test the performance of microbiological assays
Culture media and base ingredients are conveniently organized by food and beverage application for ease of locating the proper materials.
Visit our Microbial Culture Media and Microbiology Standards webpages for more information.
Culture Media for Pathogen Detection
For simple and fast detection of bacteria, use media with a chromogenic substrate such as Salmon-GAL, X-Gal, and X-glucuronide. Certain enzymes, produced by some bacteria, cleave these substrates, resulting in the different coloration of specific bacteria colonies. This allows us to clearly see the presence of the targeted microorganism, observed as a clearly visible color change. Most chromogenic media is intended for the detection of pathogenic bacteria, including our media for Clostridum perfringens, the first of its kind.
- Media for Brewery, Winery, and Fermentations
- Media for the Dairy Industry
- Disinfectants Testing
- Food Types
- Media for sterility testing
- Vitamin testing
- Media for water testing
- Media by microorganism
We offer a wide selection of tests and reagents for pathogen detection, identification, and confirmation.
Visit our Quality Indicator Testing Devices webpage for more information.
Test Kits for Classic Biochemical Methods
Classical reagents, kit systems, or convenient discs and strips can be used in the identification and confirmation of microorganisms. Based on rapid screening methods like the detection of enzymes with chromogenic substrates, indicators, or complex building reactions, they are faster and easier to use. Also, the sensitivity to certain inhibitory substances can be used to identify organisms.
- Biochemical disc and strip tests
- Biochemical reagents
Visit our Microbial Identification Tests & Biochemical Reagents webpage for more information.
Test Kits for Innovative Molecular Biological Methods
HybriScan rapid test kits are based on an rRNA sandwich hybridization system which is less expensive and more robust than PCR methods. They are also more accurate (only detects living cells), quicker (time savings of up to 10 days over cultivation-based assays) and can be performed with standard laboratory equipment. These kits can be used for the identification and quantification of spoilage and pathogenic microorganisms in foods and beverages. They are ideal for the comprehensive and reliable routine control of raw materials and concentrate on all production steps, up to the quality check of finished goods.
Visit our Pathogen Screening Kits, Instruments & Disposables webpage for more information.
Test Kits for Innovative Immunological Methods
Our Vitroids™ and LENTICULE® discs range offers certified reference test strains under double accreditation. They are certified according to ISO 17025, produced under ISO Guide 34, and meet UKAS guidelines. These products contain a precisely defined and certified CFU level in a dried state on small discs.
We also offer certified reference DNA.
- Microbiology CRMs
- DNA certified CRMs
More resources
- Microbiology equipment
- Microscopy reagents and kits
- Food Microbiological Control