Merck
  • Novel and potent 6-chloro-3-pyridinyl ligands for the alpha4beta2 neuronal nicotinic acetylcholine receptor.
Novel and potent 6-chloro-3-pyridinyl ligands for the alpha4beta2 neuronal nicotinic acetylcholine receptor.
Journal of medicinal chemistry (1999-06-23)
B Latli, K D'Amour, J E Casida
ABSTRACT
1-[(6-Chloro-3-pyridinyl)methyl]-2-imidazolidine (1), the N-desnitro metabolite of the major insecticide imidacloprid, is known to have similar potency to that of (-)-nicotine as an inhibitor of [3H](-)-nicotine binding at the rat recombinant alpha4beta2 neuronal nicotinic acetylcholine receptor (nAChR); IC50 values in the present study are 3.8 nM for (-)-nicotine, 6.0 nM for 1, and 155 nM for imidacloprid. Synthesis of new analogues of 1, modified only in the heterocyclic moiety (five-, six-, or seven-membered rings with NH, S, O, and CH2 substituents), gave compounds varying from 4-fold higher potency (2-iminothiazole analogue 10) to >6000-fold less active than (-)-nicotine. Other potent N-[(6-chloro-3-pyridinyl)methyl] compounds are those in which the heterocyclic imine is replaced with pyrrolidine (19) (IC50 9 nM) or trimethylammonium (22) (IC50 18 nM). A novel conversion of (-)-nicotine to its 6-chloro analogue increased the potency 2-fold. These 6-chloro-3-pyridinyl compounds are of interest as novel nAChR probes and potential metabolites of candidate insecticides.
Sigma-Aldrich
2-Chloro-5-(chloromethyl)pyridine, 97%