Merck
  • Enhanced catalase-like activity of manganese salen complexes in water: effect of a three-dimensionally fixed auxiliary.
Enhanced catalase-like activity of manganese salen complexes in water: effect of a three-dimensionally fixed auxiliary.
Chemical communications (Cambridge, England) (2006-12-01)
Yoritada Watanabe, Azusa Namba, Naoki Umezawa, Masatoshi Kawahata, Kentaro Yamaguchi, Tsunehiko Higuchi
ABSTRACT
A new Mn(Salen) complex bearing an ureido group as an auxiliary that is three-dimensionally fixed by a cyclopentane ring fused to the Salen structure was developed. This compound exhibited considerably higher catalase-like activity than the original Mn(Salen), i.e., the cyclopentane-fused Mn(Salen) without the auxiliary, under near-physiological conditions.
Sigma-Aldrich
EUK-8, ≥95% (HPLC)