Merck
Effects of fluoroquinolones on CYP4501A and 3A in male broilers.
Research in veterinary science (2010-06-29)
Ling-Ling Zhang, Jun-Ren Zhang, Kuan Guo, Hui Ji, Yan Zhang, Shan-Xiang Jiang
ABSTRACT
The inhibitory effects of fluoroquinolones on the enzyme activity, protein levels and mRNA expression of liver cytochrome P450 (CYP) 1A and 3A were investigated in male broiler chicks. Enrofloxacin (20 mg/kg), sarafloxacin (8 mg/kg) and marbofloxacin (5.5 mg/kg) were administrated in drinking water for 7 consecutive days. A cocktail of the probe drugs caffeine and dapsone was used to determine CYP1A and 3A activity. Western blot analysis and real-time PCR were used to determine the effects on protein levels of CYP1A and 3A, and on CYP1A4, 1A5, 3A37 mRNA levels. Enrofloxacin increased the half-life of elimination for both caffeine and dapsone, and decreased expression of CYP1A and 3A protein. Marbofloxacin decreased the metabolism of caffeine and expression of CYP1A protein. However, no change in mRNA expression was observed for any treatment group. This suggested that high doses of enrofloxacin and marbofloxacin, but not sarafloxacin, inhibit CYP in chick liver raising the possibility of drug-drug interaction when using these compounds.
Product Description
Supelco
Sarafloxacin hydrochloride hydrate, VETRANAL®, analytical standard