Merck
  • Time-dependent changes in hypoxia- and gliosis-related factors in experimental diabetic retinopathy.
Time-dependent changes in hypoxia- and gliosis-related factors in experimental diabetic retinopathy.
Eye (London, England) (2018-11-08)
Limin Gu, Hua Xu, Chaoyang Zhang, Qian Yang, Limei Zhang, Jingfa Zhang
ABSTRACT
Diabetes causes various biochemical changes in the retina; long-term changes in the factors associated with hypoxia and gliosis have rarely been reported. The present study was conducted to explore the changes in these factors in a time-dependent manner in experimental diabetic retinopathy (DR). Diabetes was induced in Sprague-Dawley rats by intraperitoneal injection of streptozotocin. The expression of the following factors was examined using immunofluorescence and western blot analysis at 0.5, 1, 2, 4 and 6 months after diabetes onset: hypoxia-inducible factor-1alpha (HIF-1alpha), vascular endothelial growth factor (VEGF), erythropoietin (EPO), erythropoietin receptor (EPOR), glial fibrillary acidic protein (GFAP), vimentin, glutamate-aspartate transporter (GLAST) and glutamine synthase (GS). The expression of factors such as HIF-1alpha, VEGF, EPO, EPOR, GFAP and vimentin, was up-regulated with the progression of diabetes in the diabetic rat retinas compared to the expression in normal control retinas, whereas the expression of GS and GLAST was down-regulated. Changes in EPO and EPOR appeared 2 weeks after diabetes onset. HIF-1alpha, VEGF and GFAP started to increase at 1 month and vimentin at 4 months after diabetes onset. GS and GLAST started to decrease at 1 month after diabetes onset. The expression of these factors, which are involved in the processes of hypoxia and gliosis, varied at different stages of DR. The time-course change may be helpful in the evaluation of the progression of DR, and it may indicate the optimal intervention time points for DR.
Sigma-Aldrich
Anti-Hypoxia Inducible Factor 1 α Antibody, clone H1α67, clone H1alpha67, Chemicon®, from mouse
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid