Merck
  • Tpl2 is required for VEGF-A-stimulated signal transduction and endothelial cell function.
Tpl2 is required for VEGF-A-stimulated signal transduction and endothelial cell function.
Biology open (2019-05-11)
Gareth W Fearnley, Izma Abdul-Zani, Antony M Latham, Monica C Hollstein, John E Ladbury, Stephen B Wheatcroft, Adam F Odell, Sreenivasan Ponnambalam
ABSTRACT
New blood vessel sprouting (angiogenesis) and vascular physiology are fundamental features of metazoan species but we do not fully understand how signal transduction pathways regulate diverse vascular responses. The vascular endothelial growth factor (VEGF) family bind membrane-bound receptor tyrosine kinases (VEGFRs), which trigger multiple signal transduction pathways and diverse cellular responses. We evaluated whether the MAP3K family member and proto-oncoprotein Tpl2 (MAP3K8) regulates basal and VEGF-A-stimulated signal transduction in endothelial cells. Notably, stimulation with exogenous VEGF-A increased Tpl2 mRNA levels and consequently de novo protein synthesis. Depletion of Tpl2 levels reveals a role in both basal and VEGF-A-stimulated endothelial cell responses, including endothelial-leukocyte interactions, monolayer permeability and new blood vessel formation. Under basal conditions, Tpl2 modulates a signal transduction cascade resulting in phosphorylation of a nuclear transcription factor (ATF-2) and altered endothelial gene expression, a pathway previously identified as crucial in VEGF-dependent vascular responses. Loss of Tpl2 expression or activity impairs signal transduction through Akt, eNOS and ATF-2, broadly impacting on endothelial function. Our study now provides a mechanism for Tpl2 as a central component of signal transduction pathways in the endothelium.
Sigma-Aldrich
PMA, for use in molecular biology applications, ≥99% (HPLC)
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Sigma-Aldrich
Tpl2 Kinase Inhibitor, The Tpl2 Kinase Inhibitor, also referenced under CAS 871307-18-5, controls the biological activity of Tpl2 Kinase. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.