Merck
  • Polyphenol rich botanicals used as food supplements interfere with EphA2-ephrinA1 system.
Polyphenol rich botanicals used as food supplements interfere with EphA2-ephrinA1 system.
Pharmacological research (2011-07-12)
Iftiin Hassan Mohamed, Carmine Giorgio, Renato Bruni, Lisa Flammini, Elisabetta Barocelli, Damiano Rossi, Giuseppe Domenichini, Ferruccio Poli, Massimiliano Tognolini
ABSTRACT
The Eph tyrosine kinase receptors and their ephrin ligands play a central role in several human cancers and their deregulated expression or function promotes tumorigenesis, inducing aggressive tumor phenotypes. Green tea extracts (GTE) have been recently found to inhibit Eph-kinase phosphorylation. In order to evaluate the potential contribution of edible and medicinal plants on EphA2-ephrinA1 modulation, 133 commercially available plant extracts used as food supplements, essential and fixed oils were screened with an ELISA-based binding assay. Nine plant extracts, rich of polyphenols, reversibly inhibited binding in a dose-dependent manner (IC₅₀ 0.83-24 μg/ml). Functional studies on PC3 prostate adenocarcinoma cells revealed that active extracts antagonized ephrinA1-Fc-induced EphA2-phosphorylation at non-cytotoxic concentrations (IC₅₀ 0.31-11.3 μg/ml) without interfering with EGF-induced EGFR activation, suggesting a specific effect. These findings could furnish an interesting starting point regarding the potential relationship between diet, edible plant secondary metabolites and Eph-ephrin system, suggesting their possible involvement in cancer development modulation.
Sigma-Aldrich
Human IgG, Fc Fragment