
Gestodene

Synonym(s):
(17α)-13-Ethyl-17-hydroxy-18,19-Dinorpregna-4,15-dien-20-yn-3-one, Gestinol, SHB 331, WL 70
Empirical Formula (Hill Notation):
C21H26O2
CAS Number:
Molecular Weight:
310.43
Products
Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Purity
Format
Research Area
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing