Skip to Content
Merck
Di-2-thienylglycolic acid methyl ester certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Di-2-thienylglycolic acid methyl ester

Di-2-thienylglycolic acid methyl ester certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Synonym(s):
Methyl hydroxy[di(thiophen-2-yl)]acetate
Empirical Formula (Hill Notation):
C11H10O3S2
Molecular Weight:
254.33

Products

Brand
Formula Weight
Manufacturer
Product Line
Quality Segment
Application
Special Grade