
1-(Ethoxycarbonylmethyl)-6-methoxyquinolinium bromide

Synonym(s):
(6-Methoxyquinolinio)acetic acid ethyl ester bromide, MQAE
Empirical Formula (Hill Notation):
C14H16BrNO3
CAS Number:
Molecular Weight:
326.19
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing