Key Documents
PMFXWKITMCC
Millipore
MCC Flexware Maintenance Kit
About This Item
UNSPSC Code:
41115700
packaging
1 ea of
storage condition
room temperature
storage temp.
room temp
Compare Similar Items
View Full Comparison
Show Differences
1 of 4
This Item
HPA024756HPA040134HPA053874
Quality Level
100
Quality Level
100
Quality Level
100
Quality Level
100
antibody form
affinity isolated antibody
antibody form
affinity isolated antibody
antibody form
affinity isolated antibody
antibody form
affinity isolated antibody
conjugate
unconjugated
conjugate
unconjugated
conjugate
unconjugated
conjugate
unconjugated
product line
Prestige Antibodies® Powered by Atlas Antibodies
product line
Prestige Antibodies® Powered by Atlas Antibodies
product line
Prestige Antibodies® Powered by Atlas Antibodies
product line
Prestige Antibodies® Powered by Atlas Antibodies
storage temp.
−20°C
storage temp.
−20°C
storage temp.
−20°C
storage temp.
−20°C