Merck
HomeC–H FunctionalizationEllman's Sulfinamides
Ellman's Sulfinamides
A diagram showing three molecular structures, each featuring a sulfur atom (S) bonded to an amine group (NH2) and a carbonyl group (O). The structures vary in their connectivity, illustrating different configurations of the same functional groups.
Since its introduction by Ellman in 1997 as a chiral ammonia equivalent,1 enantiopure 2-methyl-2-propanesulfinamide (tert-butanesulfinamide) has been demonstrated to be a versatile chiral auxiliary and has found extensive use both in academics and industry. Condensation of tert-butanesulfinamide with aldehydes and ketones proceeds under mild conditions and provides tert-butanesulfinyl imines in high yields. The tert-butanesulfinyl group activates these imines for the addition of many different classes of nucleophiles and serves as a powerful chiral directing group to provide products with generally high diastereoselectivity. Subsequent removal of the tert-butanesulfinyl group under mild conditions cleanly provides the amine products.
These tert-butanesulfinyl imines have been used as intermediates in the asymmetric synthesis of many versatile building blocks2 including syn- and anti- 1,2- or 1,3-amino alcohols,3,4 α-branched and α,α-dibranched amines,5 and α- or β-amino acids and esters6,7 (Scheme 1). Several researchers have taken advantage of the robust chemistry of tert-butanesulfinyl imines in the synthesis of antibiotics, biologically active compounds and other complex natural products.8 Furthermore, tert-butanesulfinyl imines have been used in the synthesis of asymmetric ligands,9 and in a few cases, appears as the chirality-bearing component.10
A chemical diagram depicting a central molecule with a sulfur atom (S) bonded to nitrogen (N) and two functional groups (R1 and R2). Surrounding this central structure are various molecular structures, each featuring different combinations of amine (NH2), hydroxyl (OH), and carbonyl (O) groups, indicating possible reactions or interactions involving the central molecule. Arrows connect the central structure to the surrounding molecules, illustrating their relationships.
Scheme 1.Asymmetric synthesis of many versatile building blocks using tert-butanesulfinyl imines
Recently, tert-butanesulfinyl imines have been employed in the synthesis of chiral heterocycles. A few groups have synthesized chiral aziridines through a common tert-butanesulfinyl imine intermediate (Scheme 2). Morton and co-workers synthesized chiral aziridines using trimethylsulfonium iodide with good yields and diastereoselectivities.11a Chemla and Ferreira reacted a racemic allenylzinc substrate with various tert-butanesulfinyl imines to achieve trans-ethynylaziridines as diastereomerically and enantiomerically pure compounds in good yields.11b
A chemical reaction diagram illustrating the transformation of a sulfur-containing compound. The diagram shows a central structure with a sulfur atom (S) bonded to nitrogen (N) and two functional groups (R). The reaction pathway includes reagents such as trimethylsulfonium (Me₃S⁺), sodium hydride (NaH), and dimethyl sulfoxide (DMSO) at 20°C. Further steps involve zinc bromide (ZnBr) and a terminal alkyne (TMS) in diethyl ether (Et₂O) at room temperature (rt), leading to the formation of a new sulfur-containing compound on the right.
Scheme 2.synthesis of chiral aziridines through a common tert-butanesulfinyl imine intermediate
Additionally, Dondas and De Kimpe devised an efficient route to pyrrolidines and piperidines using a common racemic tert-butanesulfinyl amine (Scheme 3), which is easily achieved from the sulfinyl imine by reduction with NaBH4.12 Their synthesis highlights a one-pot cascade cyclization and fragmentation, which allows for very high yields and purity of the cyclized product.
A chemical reaction scheme illustrating the interaction of two brominated compounds featuring nitrogen (N) and various functional groups (R1 and R2). The diagram shows the reactants on the left, indicating a 100% yield in a 1:1 ratio. The central structure includes a sulfur atom (S) bonded to nitrogen and an oxygen (O) group. The reaction conditions specify the use of dichloromethane (CH₂Cl₂) at 0°C for 16 hours, leading to the formation of a new compound on the right, which includes a selenium (Se) or iodine (I) substituent. The diagram notes that the final product also achieves a 100% yield with specific substituents (E and X).
Scheme 3.Pyrrolidines and piperidines using a common racemic tert-butanesulfinyl amine
Ellman’s research group has also reported the synthesis of chiral heterocycles. In an extension of their work on synthesis of 1,3-amino-alcohols,4a Ellman carried out the asymmetric syntheses of (-)-halosaline and (-)-8-epihalosaline (Scheme 4).
A chemical reaction diagram illustrating the synthesis of two compounds, epihalosaline and halosaline, from a starting molecule featuring a nitrogen (N) atom, hydroxyl (OH) groups, and a carbon chain. The diagram shows the addition of catecholborane under specific conditions (THF at -48°C) and lithium triethylborohydride (LiBEt₃H) at -10°C. The reaction proceeds through two pathways, each involving hydrogenation (H₂) with platinum oxide (PtO₂) and subsequent treatment with trifluoroacetic acid (TFA) in ethanol (EtOH) and water (H₂O). The products, (-)-epihalosaline and (-)-halosaline, are displayed on the right.
Scheme 4.Asymmetric syntheses of (-)-halosaline and (-)-8-epihalosaline
Another recent report describes the intermolecular self-condensation of chiral tert-butanesulfinyl imines in a synthesis for the pyrrolizidine alkaloid SC-53116 (Scheme 5).13 In a later report, Ellman demonstrated the facile synthesis of chiral 2-substituted pyrrolidines (Scheme 6) that proceeds with high yields and diastereoselectivities.14
A chemical reaction diagram depicting the synthesis of the compound SC-53116. The process begins with a starting molecule featuring a sulfur atom (S), nitrogen (N), and various functional groups. The first step involves the use of lithium hexamethyldisilazane (LHMD) and N,N'-dimethylpropyleneurea (DMPU) in tetrahydrofuran (THF) at -48°C for 3 hours. The reaction proceeds to a second step involving microwave heating in acetonitrile (CH₃CN) at 150°C for 15 minutes, leading to the formation of an intermediate structure. The final product, SC-53116, is shown on the bottom right, indicating the complete synthesis.
Scheme 5.Self-condensation of chiral tert-butanesulfinyl imines to form pyrrolizidine alkaloid
A chemical reaction diagram illustrating a synthetic pathway involving a starting molecule with a sulfur atom (S), nitrogen (N), and various functional groups (R1, R2). The first step shows the addition of an organomagnesium reagent (BrMg) to an intermediate compound, leading to the formation of a new structure. The subsequent reaction involves treatment with a mixture of trifluoroacetic acid (TFA) and water (H₂O) for 10 minutes, followed by the use of triethylsilane (Et₃SiH) over 24 hours. The final product, a cyclic compound featuring nitrogen (N) and R1, is displayed on the right.
Scheme 6.Facile synthesis of chiral 2-substituted pyrrolidines
We are pleased to be able to offer this versatile and useful auxiliary in both enantiomeric and racemic forms for your research.
References
1.
Liu G, Cogan DA, Ellman JA. 1997. Catalytic Asymmetric Synthesis oftert-Butanesulfinamide. Application to the Asymmetric Synthesis of Amines. J. Am. Chem. Soc.. 119(41):9913-9914. https://doi.org/10.1021/ja972012z
2.
Ellman JA, Owens TD, Tang TP. 2002. N-tert-Butanesulfinyl Imines:  Versatile Intermediates for the Asymmetric Synthesis of Amines. Acc. Chem. Res.. 35(11):984-995. https://doi.org/10.1021/ar020066u
3.
NA. NA.
4.
Evans JW, Ellman JA. 2003. Stereoselective Synthesis of 1,2-Disubstituted ?-Amino Alcohols by Nucleophilic Addition toN-tert-Butanesulfinyl ?-Alkoxyaldimines. J. Org. Chem.. 68(26):9948-9957. https://doi.org/10.1021/jo035224p
5.
Tang TP, Volkman SK, Ellman JA. 2001. Asymmetric Synthesis of Protected 1,2-Amino Alcohols Usingtert-Butanesulfinyl Aldimines and Ketimines. J. Org. Chem.. 66(26):8772-8778. https://doi.org/10.1021/jo0156868
6.
Barrow JC, Ngo PL, Pellicore JM, Selnick HG, Nantermet PG. 2001. A facile three-step synthesis of 1,2-amino alcohols using the Ellman homochiral tert-butylsulfinamide. Tetrahedron Letters. 42(11):2051-2054. https://doi.org/10.1016/s0040-4039(01)00122-8
7.
Kochi T, Tang TP, Ellman JA. 2003. Development and Application of a New General Method for the Asymmetric Synthesis ofsyn- andanti-1,3-Amino Alcohols. J. Am. Chem. Soc.. 125(37):11276-11282. https://doi.org/10.1021/ja0363462
8.
Kochi T, Tang TP, Ellman JA. 2002. Asymmetric Synthesis ofsyn- andanti-1,3-Amino Alcohols. J. Am. Chem. Soc.. 124(23):6518-6519. https://doi.org/10.1021/ja026292g
9.
Cogan DA, Liu G, Ellman J. 1999. Asymmetric synthesis of chiral amines by highly diastereoselective 1,2-additions of organometallic reagents to N-tert-butanesulfinyl imines. Tetrahedron. 55(29):8883-8904. https://doi.org/10.1016/s0040-4020(99)00451-2
10.
Cogan DA, Ellman JA. 1999. Asymmetric Synthesis of ?,?-Dibranched Amines by the Trimethylaluminum-Mediated 1,2-Addition of Organolithiums totert-Butanesulfinyl Ketimines. J. Am. Chem. Soc.. 121(1):268-269. https://doi.org/10.1021/ja983217q
11.
Avenoza A, Peregrina J, Busto J, Corzana F, Sucunza D, Zurbano M. Diastereoselective Synthesis of (S)- and (R)-?-Phenylserine by a Sulfinimine-Mediated Strecker Reaction. Synthesis. 2005(04):575-578. https://doi.org/10.1055/s-2004-837308
12.
Naskar D, Roy A, Seibel WL, Portlock DE. 2003. Hydroxylamines and sulfinamide as amine components in the Petasis boronic acid?Mannich reaction: synthesis of N-hydroxy or alkoxy-?-aminocarboxylicacids and N-(tert-butyl sulfinyl)-?-amino carboxylicacids. Tetrahedron Letters. 44(49):8865-8868. https://doi.org/10.1016/j.tetlet.2003.09.179
13.
Jacobsen MF, Skrydstrup T. 2003. Asymmetric Mannich-Type Reactions for the Synthesis of Aspartic Acid Derivatives from ChiralN-tert-Butanesulfinylimino Esters. J. Org. Chem.. 68(18):7112-7114. https://doi.org/10.1021/jo034436j
14.
Tang TP, Ellman JA. 2002. Asymmetric Synthesis of ?-Amino Acid Derivatives Incorporating a Broad Range of Substitution Patterns by Enolate Additions totert-Butanesulfinyl Imines. J. Org. Chem.. 67(22):7819-7832. https://doi.org/10.1021/jo025957u
15.
Tang TP, Ellman JA. 1999. Thetert-Butanesulfinyl Group:  An Ideal Chiral Directing Group and Boc-Surrogate for the Asymmetric Synthesis and Applications of ?-Amino Acids. J. Org. Chem.. 64(1):12-13. https://doi.org/10.1021/jo9820824
16.
Lu BZ, Senanayake C, Li N, Han Z, Bakale RP, Wald SA. 2005. Control of Diastereoselectivity by Solvent Effects in the Addition of Grignard Reagents to Enantiopuret-Butylsulfinimine:? Syntheses of the Stereoisomers of the Hydroxyl Derivatives of Sibutramine. Org. Lett.. 7(13):2599-2602. https://doi.org/10.1021/ol0507017
17.
Kochi T, Ellman JA. 2004. Asymmetric ?-Alkylation ofN?-tert-Butanesulfinyl Amidines. Application to the Total Synthesis of (6R,7S)-7-Amino-7,8-dihydro-?-bisabolene. J. Am. Chem. Soc.. 126(48):15652-15653. https://doi.org/10.1021/ja044753n
18.
Kochi T, Ellman JA. 2004. Asymmetric ?-Alkylation ofN?-tert-Butanesulfinyl Amidines. Application to the Total Synthesis of (6R,7S)-7-Amino-7,8-dihydro-?-bisabolene. J. Am. Chem. Soc.. 126(48):15652-15653. https://doi.org/10.1021/ja044753n
19.
Kato T, Marubayashi K, Takizawa S, Sasai H. 2004. Development of a novel chiral spiro ligand bearing oxazoline. Tetrahedron: Asymmetry. 15(23):3693-3697. https://doi.org/10.1016/j.tetasy.2004.09.037
20.
Schenkel LB, Ellman JA. 2004. Application ofP,N-Sulfinyl Imine Ligands to Iridium-Catalyzed Asymmetric Hydrogenation of Olefins. J. Org. Chem.. 69(6):1800-1802. https://doi.org/10.1021/jo035675+
21.
Schenkel LB, Ellman JA. 2003. Novel Sulfinyl Imine Ligands for Asymmetric Catalysis. Org. Lett.. 5(4):545-548. https://doi.org/10.1021/ol027468m
22.
Owens TD, Souers AJ, Ellman JA. 2003. The Preparation and Utility of Bis(sulfinyl)imidoamidine Ligands for the Copper-Catalyzed Diels?Alder Reaction. J. Org. Chem.. 68(1):3-10. https://doi.org/10.1021/jo020524c
23.
Chemla F, Ferreira F. 2004. High Kinetic Resolution in the Addition of a Racemic Allenylzinc onto EnantiopureN-tert-Butanesulfinimines:  Concise Synthesis of Enantiopuretrans-2-Ethynylaziridines1. J. Org. Chem.. 69(24):8244-8250. https://doi.org/10.1021/jo0490696
24.
Dondas HA, De Kimpe N. 2005. A sequence of electrophile induced cyclisation and concomitant N-deprotection of alkenylsulfinimines and alkenylsulfinamides as a direct route to cyclic or spirocyclic imines, pyrrolidines and piperidines. Tetrahedron Letters. 46(24):4179-4182. https://doi.org/10.1016/j.tetlet.2005.04.060
25.
Schenkel LB, Ellman JA. 2004. Self-Condensation ofN-tert-Butanesulfinyl Aldimines:? Application to the Rapid Asymmetric Synthesis of Biologically Important Amine-Containing Compounds. Org. Lett.. 6(20):3621-3624. https://doi.org/10.1021/ol048458j
26.
Brinner KM, Ellman JA. 2005. A rapid and general method for the asymmetric synthesis of 2-substituted pyrrolidines using tert-butanesulfinamide. Org. Biomol. Chem.. 3(11):2109. https://doi.org/10.1039/b502080h