Merck
Oxidative stress-mediated p53/p21
Chemosphere (2017-12-19)
Junguo Ma, Yuanyuan Li, Mengli Wu, Xiaoyu Li
ABSTRACT
A previous study showed that microcystin-LR (MC-LR) exerted cytotoxicity and induced apoptosis in HepG2 cells. In the present study, we investigated whether oxidative stress-mediated p53/p21