Merck
  • Caryophyllane sesquiterpenes inhibit DNA-damage by tobacco smoke in bacterial and mammalian cells.
Caryophyllane sesquiterpenes inhibit DNA-damage by tobacco smoke in bacterial and mammalian cells.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2017-11-21)
Silvia Di Giacomo, Lorena Abete, Rossana Cocchiola, Gabriela Mazzanti, Margherita Eufemi, Antonella Di Sotto
ABSTRACT
In the present study, the ability of the natural sesquiterpene β-caryophyllene (CRY) and its metabolite β-caryophyllene oxide (CRYO) to inhibit the genotoxicity of a condensate of cigarette smoke (CSC) was evaluated both in bacterial and mammalian cells. Also, the inhibition of the CSC-mediated STAT3 phosphorylation and intracellular oxidative stress was evaluated as potential chemopreventive mechanism. Under our experimental conditions, both the sesquiterpenes exhibited antimutagenic properties, being CRY the most potent compound. The antimutagenicity was highlighted in all experimental protocols, being particularly strong in the co- and post-treatments. The test substances also reduced the micronuclei frequency induced by CSC, with a major effectiveness of CRY. CRY was also able to reduce the CSC-mediated increase of the Y705- pSTAT3 levels, in spite of a lacking effect of CRYO. Furthermore, the sesquiterpenes CRY and CRYO displayed a moderate antioxidant activity, with a 25 % and 40 % inhibition of the ROS-levels increased by CSC, respectively. On the basis of these results, CRY seems to be a multi-target chemopreventive agent, although the genoprotective and antioxidant effects of CRYO suggest that both compounds deserve to be deeply investigated for a possible application in the prevention and treatment of different smoke-related ailments.
Sigma-Aldrich
2-Aminofluorene, 98%