Merck
CD34
Oncotarget (2018-02-09)
Willie Lin, Yogi Chang-Yo Hsuan, Yu-Chin Su, Cheng-Hsien Lin, Mao-Tsun Lin, Zi-Hao Chen, Ching-Ping Chang, Kao-Chang Lin
ABSTRACT
CD34 is a transmembrane phosphoglycoprotein used to selectively enrich bone marrow in hematopoietic stem cells for transplantation. Treating rats with CD34