Merck
  • Dehydropropylpantothenamide isolated by a co-culture of Nocardia tenerifensis IFM 10554
Dehydropropylpantothenamide isolated by a co-culture of Nocardia tenerifensis IFM 10554
Journal of natural medicines (2017-12-07)
Yasumasa Hara, Midori A Arai, Kanae Sakai, Naoki Ishikawa, Tohru Gonoi, Takashi Yaguchi, Masami Ishibashi
ABSTRACT
A new amide, named dehydropropylpantothenamide (1), was obtained by a co-culture of Nocardia tenerifensis IFM 10554